Please login to your online account to display your discounted pricing
Dynalon™ Azlon™ Carboys and Large Bottles
Range of high-performance large bottles and carboys.
Manufacturer: dynalon™  505634-2
 View more versions of this product
Catalog No. S01989A
  • / Each
Description & Specifications
Specifications
Product Type Carboy
Capacity (English) 2.64 gal.
Capacity (Metric) 10L
Shape Rounded octagonal
Handles Blow molded
Capacity 2.64 gal., 10L
Neck Style Offset
Material Polypropylene
Autoclavable Yes