Please login to your online account to display your discounted pricing
Dynalon™ Azlon™ Carboys and Large Bottles
Range of high-performance large bottles and carboys.
$101.35 - $171.10
Specifications
Autoclavable Yes
Handles Blow molded
Material Polypropylene
Neck Style Offset
Shape Rounded octagonal
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity    
S01989
dynalon™
505634-1
1.32 gal. 5L Each for $101.35
S01989B
dynalon™
505634-3
5.28 gal. 20L Each for $171.10
S01989A
dynalon™
505634-2
2.64 gal. 10L Each for $142.35
Description & Specifications
Specifications
Autoclavable Yes
Handles Blow molded
Material Polypropylene
Neck Style Offset
Shape Rounded octagonal
  • Autoclavable polypropylene™, heavy-duty construction
  • Stylish rounded octagonal shape for increased rigidity when full
  • Offset neck for easier filling and pouring
  • Integral, comfortable blow molded handles for ease of carrying, are end-sealed to prevent contamination
  • Heavy-duty closures
  • Carboys feature a durable and leakproof heavy-duty spigot with boss fitting for extra strength
  • Spigot features removable tube fitting for added convenience
  • Molded graduations allow easy viewing of contents