Please login to your online account to display your discounted pricing

Alfa Aesar™ Azithromycin dihydrate, 98%


Quantity 5g

Manufacturer: alfa aesar™  J6674006

 View more versions of this product

Catalog No. AAJ6674006

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 785.01
CAS 117772-70-0
Chemical Name or Material Azithromycin dihydrate
MDL Number MFCD01862248
Assay 98%
Physical Form Powder
Synonym N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A; CP-62993
Melting Point 126°C
Boiling Point 717°C
Color White
Solubility Information Soluble in DMSO at 20mg/mL; Also soluble in chloroform or ethylene chloride
RTECS Number RN6960000
Storage Note 1 2° to 8°C
Quantity 5g
Sensitivity Air Sensitive

Specifications

Chemical Name or Material Azithromycin dihydrate
CAS 117772-70-0
Formula Weight 785.01
MDL Number MFCD01862248
Synonym N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A; CP-62993

Applications
Azithromycin dihydrate has anti-immunomodulatory/anti-inflammatory properties, which make it useful in treating cystic fibrosis. It shows an azalide antimicrobial agent active in vitro against various pathogens.

Solubility
Soluble in DMSO at 20mg/ml. Also soluble in chloroform or ethylene chloride

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Accessories

2° to 8°C

SDS & Safety Documents