missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azidotrimethylsilane, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC167932500

 View more versions of this product

Catalog No. AC167932500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

C-X bond formation reagent, For Azidation/Diazo transfer
Specifications

Specifications

Azidotrimethylsilane
Authentic
97%
Glass bottle
01,1236; 03,316; 05,719; 06,632; 07,394; 09,21; 10,14; 11,32; 12,37; 13,24; 14,25; 15,16; 16,17; 17,378
Trimethylsilyl azide
SEDZOYHHAIAQIW-UHFFFAOYSA-N
azido(trimethyl)silane
78378
Liquid
Colorless through Yellow
250mL
0.876
4648-54-8
97%
C3H9N3Si
(CH3)3SiN3
1.4142 to 1.4162
Solubility in water: decomposes. Other solubilities: soluble in ether, dichloromethane and benzene
C[Si](C)(C)N=[N+]=[N-]
115.211
115.21
0.8760g/mL
95.0°C to 96.0°C
6°C

Chemical Identifiers

4648-54-8
115.211
Trimethylsilyl azide
C3H9N3Si
SEDZOYHHAIAQIW-UHFFFAOYSA-N
78378
Safety and Handling

Safety and Handling

Danger

SDS Documents