missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Azidotri-n-butyltin(IV), 95%

Manufacturer:  Alfa Aesar™ H5563414

 View more versions of this product

Catalog No. AAH5563414



Description

Description

Applications
Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.

Solubility
Miscible with toluene, hexane, acetonitrile and tetrahydrofuran.

Notes
Use in a fume hood. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Azidotri-n-butyltin(IV)
113°C (235°F)
17846-68-3
[CH3(CH2)3]3SnN3
UN2788
Miscible with toluene,hexane,acetonitrile and tetrahydrofuran.
CCCC[Sn](CCCC)(CCCC)N=[N+]=[N-]
332.079
332.06
1.212
25g
C12H27N3Sn
MFCD00216557
azido tributyl stannane
JKVRTUCVPZTEQZ-UHFFFAOYSA-N
azido(tributyl)stannane
4984872
95%

Chemical Identifiers

17846-68-3
332.079
JKVRTUCVPZTEQZ-UHFFFAOYSA-N
4984872
CCCC[Sn](CCCC)(CCCC)N=[N+]=[N-]
C12H27N3Sn
MFCD00216557
azido tributyl stannane
azido(tributyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents