missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azidotri-n-butyltin(IV), 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5563406

 View more versions of this product

Catalog No. AAH5563406



Description

Description

Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.

Solubility
Miscible with toluene, hexane, acetonitrile and tetrahydrofuran.

Notes
Use in a fume hood. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Azidotri-n-butyltin(IV)
1.212
C12H27N3Sn
MFCD00216557
UN2788
Miscible with toluene,hexane,acetonitrile and tetrahydrofuran.
CCCC[Sn](CCCC)(CCCC)N=[N+]=[N-]
332.079
332.06
17846-68-3
113°C (235°F)
[CH3(CH2)3]3SnN3
5 g
tributyltin azide, azidotributyltin iv, azidotributyltin, azidotributyl tin, tri-n-butylazidotin, azido tributyl stannane, azido tributyltin, tri-butyltin azide, tributyl tin azide, tributylstannylazide
JKVRTUCVPZTEQZ-UHFFFAOYSA-N
azido(tributyl)stannane
4984872
95%

Chemical Identifiers

17846-68-3
332.079
JKVRTUCVPZTEQZ-UHFFFAOYSA-N
4984872
CCCC[Sn](CCCC)(CCCC)N=[N+]=[N-]
C12H27N3Sn
MFCD00216557
tributyltin azide, azidotributyltin iv, azidotributyltin, azidotributyl tin, tri-n-butylazidotin, azido tributyl stannane, azido tributyltin, tri-butyltin azide, tributyl tin azide, tributylstannylazide
azido(tributyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only