Please login to your online account to display your discounted pricing

Alfa Aesar™ Azido-PEG3-carboxytetramethylrhodamine 110 conjugate

Chemical Identifiers

Chemical Name or Material Azido-PEG3-carboxytetramethylrhodamine 110 conjugate
Molecular Formula C33H38N6O7
Formula Weight 630.7
MDL Number MFCD22380734
Synonym Azide-Fluor 545
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64510LB0

  SDS
alfa aesar™
J64510LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 630.7
MDL Number MFCD22380734
Molecular Formula C33H38N6O7
Chemical Name or Material Azido-PEG3-carboxytetramethylrhodamine 110 conjugate
Recommended Storage −30° to −10°C
Synonym Azide-Fluor 545
Quantity 5mg
Solubility Information Soluble in water
Color Dark Red
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Physical Form Solid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Azido-PEG3-carboxytetramethylrhodamine is used as a water soluble azide functionalized- TAMRA dye. The water-soluble PEG spacer minimizes the interaction between the dye and the biomolecule to be labeled, used to detect or label alkyne- or cyclooctyne-containing molecules or biomolecules by fluorescence spectroscopy following an azide-alkyne click chemistry reaction.

Solubility
Soluble in water

Notes
Absorbance maximum at 546nm; maximum emission at 565nm. Extinction coefficient: 92,000 per cm per M.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C