missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Azido-PEG3-biotin conjugate

An azide-biotin conjugate with PEG spacer to improve solubility in aqueous media

Manufacturer:  Alfa Aesar™ J64996MC

 View more versions of this product

Catalog No. AAJ64996MC



Description

Description

Applications
Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it′s components do not react with the biological environment.

Solubility
Solubile in DMSO, DMF and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Azido-PEG3-biotin conjugate
100mg
C18H32N6O5S
11-r-+-biotinylamino-1-azido-3,6,9-trioxaundecane
ZWFOOMQCYIGZBE-ZOBUZTSGSA-N
5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]pentanamide
60146223
Solid
White to Gray
875770-34-6
MFCD20134145
Soluble in DMSO, DMF and water
C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCN=[N+]=[N-])NC(=O)N2
444.551
444.55

Chemical Identifiers

875770-34-6
444.551
ZWFOOMQCYIGZBE-ZOBUZTSGSA-N
60146223
C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCN=[N+]=[N-])NC(=O)N2
C18H32N6O5S
MFCD20134145
11-r-+-biotinylamino-1-azido-3,6,9-trioxaundecane
5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]pentanamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C

Documents