missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Azido-PEG11-biotin conjugate

An azide-biotin conjugate with a long PEG spacer

Chemical Identifiers

Molecular Formula C34H64N6O13S
MDL Number MFCD21363313
Synonym 35-[(R)-(+)-Biotinylamino]-1-azido-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontane
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65147MB View Documents Alfa Aesar™
J65147MB
25mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ65147MC View Documents Alfa Aesar™
J65147MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
An azide-biotin conjugate with a long PEG spacer, Biotinylation reagent for labeling alkyne containing molecules or biomolecules. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest.

Solubility
Soluble in DMSO, DMF and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

C34H64N6O13S
MFCD21363313
35-[(R)-(+)-Biotinylamino]-1-azido-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontane

Specifications

White to Gray
C34H64N6O13S
35-[(R)-(+)-Biotinylamino]-1-azido-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontane
Solid
25mg
MFCD21363313
796.96
Azido-PEG11-biotin conjugate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C