missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azetidine hydrochloride, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC398730050

 View more versions of this product

Catalog No. AC398730050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Azetidine hydrochloride
5g
97%
MFCD00191762
HGQULGDOROIPJN-UHFFFAOYSA-N
azetidine;hydrochloride
12308726
Glass bottle
36520-39-5
C3H8ClN
acmc-1aiho
C1CNC1.Cl
93.554
93.56

Chemical Identifiers

36520-39-5
93.554
HGQULGDOROIPJN-UHFFFAOYSA-N
12308726
C1CNC1.Cl
C3H8ClN
MFCD00191762
acmc-1aiho
azetidine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents