Please login to your online account to display your discounted pricing

AZD 5438, Tocris™

Potent cyclin-dependent kinase (cdk) 1, 2 and 9 inhibitor

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material AZD 5438
Assay Percent Range >99%
Molecular Formula C18H21N5O2S
Formula Weight 371.46
Synonym 4-[2-Methyl-1-(1-methylethyl)-1H-imidazol-5-yl]-N-[4-(methylsulfonyl)phenyl]-2-pyrimidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

396850

 
r & d systems
396850
50mg Each for $1,180.32

396810

 
r & d systems
396810
10mg Each for $284.81
Description & Specifications

Specifications

Chemical Name or Material AZD 5438
CAS 602306-29-6
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >99%
Molecular Formula C18H21N5O2S
Formula Weight 371.46
Synonym 4-[2-Methyl-1-(1-methylethyl)-1H-imidazol-5-yl]-N-[4-(methylsulfonyl)phenyl]-2-pyrimidinamine
Quantity 50mg
Safety and Handling

Desiccate at Room Temperature