missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Azadibenzocyclooctyne-S-S-PEG3-biotin conjugate

Chemical Identifiers

Molecular Formula C42H56N6O8S3
MDL Number MFCD22380733
Synonym ADIBO-S-S-PEG3-biotin conjugate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64256MA View Documents Alfa Aesar™
J64256MA
10mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ64256MB View Documents Alfa Aesar™
J64256MB
25mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • A cleavable biotinylation reagent with a PEG spacer
  • The PEG spacer allows for more efficient binding, availability and solubility
Specifications

Chemical Identifiers

C42H56N6O8S3
MFCD22380733
ADIBO-S-S-PEG3-biotin conjugate

Specifications

Gray to Yellow
C42H56N6O8S3
ADIBO-S-S-PEG3-biotin conjugate
Solid
10mg
MFCD22380733
869.12
Azadibenzocyclooctyne-S-S-PEG3-biotin conjugate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C