missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Azadibenzocyclooctyne-PEG4-NHS ester

Chemical Identifiers

MDL Number MFCD22380747
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64126MA
View Documents
Alfa Aesar™
J64126MA
10mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ64126MB
View Documents
Alfa Aesar™
J64126MB
25mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ64126MC
View Documents
Alfa Aesar™
J64126MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.

Solubility
Soluble in DCM, DMF, DMSO and THF.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

MFCD22380747

Specifications

Moisture sensitive
10mg
Soluble in DCM,DMF,DMSO and THF.
Oil
Orange to Brown
MFCD22380747
649.69
Azadibenzocyclooctyne-PEG4-NHS ester
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C