missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Azadibenzocyclooctyne-PEG4-NHS ester

Manufacturer:  Alfa Aesar™ J64126MC

 View more versions of this product

Catalog No. AAJ64126MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.

Solubility
Soluble in DCM, DMF, DMSO and THF.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications

Specifications

Azadibenzocyclooctyne-PEG4-NHS ester
Orange to Brown
MFCD22380747
649.69
Moisture sensitive
100mg
Soluble in DCM,DMF,DMSO and THF.
Oil

Chemical Identifiers

MFCD22380747
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents