missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Azadibenzocyclooctyne-maleimide, Thermo Scientific™
For incorporation of ADIBO functionality into sulfhydryl containing compounds
Chemical Identifiers
Molecular Formula | C25H21N3O4 |
---|---|
MDL Number | MFCD22380750 |
Synonym | ADIBO-maleimide |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ65377MB
|
Thermo Scientific Chemicals
J65377MB |
25mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AAJ65377MC
|
Thermo Scientific Chemicals
J65377MC |
100mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
C25H21N3O4 | |
MFCD22380750 | |
ADIBO-maleimide |
Specifications
C25H21N3O4 | |
25mg | |
Solulbe in dichloromethane,DMF,DMSO,or THF | |
Azadibenzocyclooctyne-maleimide |
MFCD22380750 | |
ADIBO-maleimide | |
427.45 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO â Research Use Only