Learn More
Azadibenzocyclooctyne-amine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J65637MB
Description
Azadibenzocyclooctyne amine is a carbonyl reactive reagent used to incorporate ADIBO into organic compounds, surfaces or particlesAzadibenzocyclooctyne amine is widely useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAzadibenzocyclooctyne amine is a carbonyl reactive reagent used to incorporate ADIBO into organic compounds, surfaces or particlesAzadibenzocyclooctyne amine is widely useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.
Notes
Heat sensitive. Incompatible with amines and azides.
Specifications
Azadibenzocyclooctyne-amine | |
Yellow to Orange | |
MFCD22380759 | |
OCCYFTDHSHTFER-UHFFFAOYSA-N | |
276.339 | |
276.33 | |
Solid |
1255942-06-3 | |
C18H16N2O | |
dbco-amine, azadibenzocyclooctyne-amine, adibo-ch2 2-nh2.tfa, azadibenzocyclooctyne-ch2 2-amine.tfa, dibenzocyclooctyne-amine, for copper-free click chemistry, 5-beta-alanyl-11,12-didehydro-5,6-dihydrodibenzo b,f azocine, 3-amino-1-5-aza-3,4:7,8-dibenzocyclooct-1-yne-5-yl-1-propanone, 3-amino-1-2-azatricyclo 10.4.0.0?,? hexadeca-1 16 ,4,6,8,12,14-hexaen-10-yn-2-yl propan-1-one, 3-amino-1-2-azatricyclo 10.4.0.04,9 hexadeca-1 12 ,4 9 ,5,7,13,15-hexaen-10-yn-2-yl propan-1-one | |
C1C2=CC=CC=C2C#CC3=CC=CC=C3N1C(=O)CCN | |
77078258 | |
25 mg |
Chemical Identifiers
1255942-06-3 | |
276.339 | |
OCCYFTDHSHTFER-UHFFFAOYSA-N | |
77078258 |
C18H16N2O | |
MFCD22380759 | |
dbco-amine, azadibenzocyclooctyne-amine, adibo-ch2 2-nh2.tfa, azadibenzocyclooctyne-ch2 2-amine.tfa, dibenzocyclooctyne-amine, for copper-free click chemistry, 5-beta-alanyl-11,12-didehydro-5,6-dihydrodibenzo b,f azocine, 3-amino-1-5-aza-3,4:7,8-dibenzocyclooct-1-yne-5-yl-1-propanone, 3-amino-1-2-azatricyclo 10.4.0.0?,? hexadeca-1 16 ,4,6,8,12,14-hexaen-10-yn-2-yl propan-1-one, 3-amino-1-2-azatricyclo 10.4.0.04,9 hexadeca-1 12 ,4 9 ,5,7,13,15-hexaen-10-yn-2-yl propan-1-one | |
C1C2=CC=CC=C2C#CC3=CC=CC=C3N1C(=O)CCN |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only