Please login to your online account to display your discounted pricing

AZ 3146, Tocris™

Potent and selective monopolar spindle 1 (Mps1) kinase inhibitor

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material AZ 3146
Assay Percent Range >99%
Molecular Formula C24H32N6O3
Formula Weight 452.55
Synonym 9-Cyclopentyl-2-[[2-methoxy-4-[(1-methylpiperidin-4-yl)oxy]-phenyl]amino]-7-methyl-7, 9-dihydro-8H-purin-8-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

399450

 
r & d systems
399450
50mg Each for $1,180.32

399410

 
r & d systems
399410
10mg Each for $284.81
Description & Specifications

Specifications

Chemical Name or Material AZ 3146
Quantity 50mg
CAS 1124329-14-1
Recommended Storage Store at +4°C
Assay Percent Range >99%
Molecular Formula C24H32N6O3
Formula Weight 452.55
Synonym 9-Cyclopentyl-2-[[2-methoxy-4-[(1-methylpiperidin-4-yl)oxy]-phenyl]amino]-7-methyl-7, 9-dihydro-8H-purin-8-one
Safety and Handling

Store at +4°C