missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AZ 12080282 dihydrochloride, Tocris™

Inhibitor of Hedgehog (Hh) signaling

$283.92 - $1,179.73

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C23H20N4O2·2HCl
Formula Weight 457.35
Synonym N-[5-(1H-Imidazol-2-yl)-2-methylphenyl]-4-(2-pyridinylmethoxy)benzamide dihydrochloride
Chemical Name or Material AZ 12080282 dihydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
483850 R & D Systems
RDS483850
50mg Each for $1,179.73
483810 R & D Systems
RDS483810
10mg Each for $283.92
Specifications Description & Specifications

Chemical Identifiers

>98%
457.35
AZ 12080282 dihydrochloride
C23H20N4O2·2HCl
N-[5-(1H-Imidazol-2-yl)-2-methylphenyl]-4-(2-pyridinylmethoxy)benzamide dihydrochloride

Specifications

>98%
457.35
50mg
C23H20N4O2·2HCl
N-[5-(1H-Imidazol-2-yl)-2-methylphenyl]-4-(2-pyridinylmethoxy)benzamide dihydrochloride
AZ 12080282 dihydrochloride

Safety and Handling

Recommended Storage : Desiccate at Room Temperature