Please login to your online account to display your discounted pricing

AZ 10606120 dihydrochloride, Tocris

Potent P2X7 receptor antagonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material AZ 10606120 dihydrochloride
Assay Percent Range >98%
Molecular Formula C25H34N4O2·2HCl
Formula Weight 495.48
Synonym N-[2-[[2-[(2-Hydroxyethyl)amino]ethyl]amino]-5-quinolinyl]-2-tricyclo[3.3.1.13, 7]dec-1-ylacetamide dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

332310

 
r & d systems
RDS332310
10mg Each for $214.12

332350

 
r & d systems
RDS332350
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 495.48
Quantity 10mg
CAS 607378-18-7
Synonym N-[2-[[2-[(2-Hydroxyethyl)amino]ethyl]amino]-5-quinolinyl]-2-tricyclo[3.3.1.13, 7]dec-1-ylacetamide dihydrochloride
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >98%
Molecular Formula C25H34N4O2·2HCl
Chemical Name or Material AZ 10606120 dihydrochloride
Safety and Handling

Desiccate at Room Temperature