missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ AZ 10417808

Selective non-peptide caspase-3 inhibitor

$257.85 - $1,073.10

Specifications

Product Type AZ 10417808
Inhibitors AZ 10417808
Molecular Weight 434.23
Purity >98
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
217210 Tocris™
217210
10mg Each for $257.85
217250 Tocris™
217250
50mg Each for $1,073.10
Specifications

Description

Selective non-peptide inhibitor of caspase-3 (Ki = 247nM); displays > 40-fold selectivity over caspases 1, 2, 6, 7 and 8 (Ki > 10μM). Completely blocks staurosporine-induced intracellular DEVDase activity in SH-SY5Y cells (IC50 = 14.9μM).
  • Chemical Name: 2-[(3,4-Dichlorophenyl)amino]-1,4-dihydro-6-nitro-4-oxo-N-2-propenyl-8-quinazolinecarboxamide
  • Formula: C18H13Cl2N5O4
  • CAS Number: 331645-84-2
  • Solubility: Soluble to 25mM in DMSO
  • Purity: >98
Description & Specifications

Specifications

AZ 10417808
AZ 10417808
434.23
>98