Please login to your online account to display your discounted pricing

Axygen™ Automation Tips for Zymark™/Caliper, 25µL

Compatible with automated liquid handling systems. Axygen™ Automation Tips for Zymark™/Caliper, 25µL are non-filtered pipette tips in 384-well racks. Constructed of clear polypropyle under strict controls to ensure quality. Options include sterile and non-sterile tips.

$8,778.41 - $8,778.41

Specifications

For Use With (Equipment) Zymark™/Caliper RapidPlate™, SciClone™ and Zephyr™ LV96, LV384 head
Certifications/Compliance RNase-/DNase-free
Volume (Metric) 30μL
Material Polypropylene
Shape Round
View More Specs
Products
Catalog Number Mfr. No. Sterility Price Quantity    

14-222-915

 
axygen™
ZT384CR
Nonsterile Case of 19200 Each for $8,778.41
Description & Specifications

Specifications

For Use With (Equipment) Zymark™/Caliper RapidPlate™, SciClone™ and Zephyr™ LV96, LV384 head
Certifications/Compliance RNase-/DNase-free
Volume (Metric) 30μL
Material Polypropylene
Shape Round
Filtered No
Color Clear
For Use With (Application) Non-pyrogenic

  • Comprehensive line of pipette tips for automated liquid handling systems
  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Compatible with Zymark™/Caliper RapidPlate™, SciClone™ and Zephyr™ with LV96 and LV384 head

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case