missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ Wellpro™ Robotic Tips

Clear tips

$1,553.88 - $1,654.04

Specifications

Clean Claims DNase, Rnase Free
Color Clear
For Use With (Equipment) ProGroup Wellpro™ 3000
Material PP (Polypropylene)
Tip Style Automation
View More Specs

Products 2
Catalog Number Mfr. No. Sterility Price Quantity  
Catalog Number Mfr. No. Sterility Price Quantity  
14-222-863 View Documents Corning Incorporated
TWP200RS
Sterile Case of 4800 Each for $1,654.04
 
14-222-862 View Documents Corning Incorporated
TWP200R
Non-sterile Case of 4800 Each for $1,553.88
 
Description

Description

  • Racked
  • 200μL capacity
  • Presterilized tips available
  • Grey racks

Order Info

30++l
200++l

Specifications

Specifications

DNase, Rnase Free
ProGroup Wellpro™ 3000
Automation
Non-pyrogenic, used for stringent specifications under strict process controls
Round
Clear
PP (Polypropylene)
200μL
Gray
Rack