Axygen™ Speciality Tips
For use with Pipetman™, Costar, Onepette, 8 and 12 Pette, Eppendorf™, Finnpipette™, Labpette, 8 and 12-channel, Nichiryo, Oxford-Benchmate, Rainin™ EDP2, SocorexTitertek™, Digita
Catalog Number | Mfr. No. | Tip Style | Volume (Metric) | Sterility | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Tip Style | Volume (Metric) | Sterility | Price | Quantity | |||
14-222-082
|
Corning Incorporated
FPT384R |
Microvolume | 5 to 50 μL | Non-sterile |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Volume: 5 to 50uL
- Clear, racked tips
- Sterile or nonsterile
Order Info
Microvolume tips:
Racked: 384 tips/rack, 10 racks/pack, 5 packs/case
Macrovolume tips:
Bulk packaging: 250 tips/pack, 10 packs/case
Specifications
Autoclavable | |
PP (Polypropylene) | |
Clear |