Axygen™ Screw Cap Tubes without Caps: Conical with Conical Bottoms
Conical tubes
Supplier: Axygen™ ST200CS
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
Specifications
RNase, DNase, pyrogen-free | |
Clear | |
Bag | |
Not Included |
2 mL | |
Polypropylene | |
Sterile | |
Conical |