Axygen™ PerkinElmer™/Packard Robotic Tips: Maxymum Recovery
Ideal for MiniTrak, PlateTrak and Evolution P-3
$837.56 - $837.56
Specifications
Certifications/Compliance RNase, DNase-free, ANSI
Color Clear
For Use With (Equipment) PerkinElmer™ Packard™ Janus™/Evolution P3™/MiniTrak™/PlateTrak™ automated liquid handling systems and MDT-96 P235, MDT-96 P50 and MDT-384 30μL head
Material Polypropylene
Rack Color Green
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Tip Style Sterility Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Product Type Tip Style Sterility Volume (Metric) Price Quantity  
50-400-674 Corning Incorporated
PK200LR
Robotic Tip Racked Nonsterile 200μL Case of 4800 Each for $837.56
Specifications
Description
Robotic tips uses no mold-releasing agents. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.
  • Tip volume: 20 to 200μL
  • Tip color: clear
  • Rack color: green
Order Info
Rack: 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Compliance
Axygen products are manufactured in a clean, controlled “state of the art” facility and certified RNase-, DNase- and endotoxin-safe
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
RNase, DNase-free, ANSI
PerkinElmer™ Packard™ Janus™/Evolution P3™/MiniTrak™/PlateTrak™ automated liquid handling systems and MDT-96 P235, MDT-96 P50 and MDT-384 30μL head
Green
Clear
Polypropylene
Round