missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ Multimek Type II Robotic Tips

Robbins-style tips for Beckman Multimek «Type II«

$1,640.63 - $1,640.63

Specifications

Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) Exposed to e-beam radiation for complete security without degrading the plastic or performance
For Use With (Equipment) Beckman Coulter™ Multimek™ Type II head
Tip Style Automation
Color Clear
View More Specs

Products 1
Catalog Number Mfr. No. Sterility Price Quantity  
Catalog Number Mfr. No. Sterility Price Quantity  
14-222-958
View Documents
Axygen™
RMT200CR
Non-sterile
Case of 4800 Each for $1,640.63
 
Description

Description

  • Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
  • Volume range: 200μL
  • Available in racked and racked sterile formats
  • Packaging: 96 tips/rack, 10 racks/pack, 5 packs/case

Specifications

Specifications

DNase, RNase, ATP, and Endotoxin Free
Beckman Coulter™ Multimek™ Type II head
Clear
200 μL
Exposed to e-beam radiation for complete security without degrading the plastic or performance
Automation
SBS Rack
SDS