missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ Multimek Robotic Tips: 50μL

Fits Multimek «Type II« or CCS-style head

$293.96 - $293.96

Specifications

Certifications/Compliance DNase, RNase, pyrogen free
Color Clear
For Use With (Equipment) Beckman™ Coulter™ MultiMek™ automated system
For Use With (Application) Unique diamond polished molding technology use no mold-releasing agents, minimizes wastage of precious samples and reagents while handling, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Tip Style Sterility Price Quantity  
Catalog Number Mfr. No. Tip Style Sterility Price Quantity  
14-222-194 Corning Incorporated
MT50LRS
Maxymum Recovery, Racked; Presterilized Sterile Pack of 960 Each for $293.96
Case of 5 Pack for $1,278.10
Specifications

Description

Robotic tip's unique diamond-polished molding technology uses no mold-releasing agents. Minimizes wastage of precious samples and reagents while handling. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.

  • Tip volume: 50 and 200μL
  • Tip color: Clear
  • Rack color: Red and blue

Order Info

Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

Description & Specifications

Specifications

DNase, RNase, pyrogen free
Clear
Beckman™ Coulter™ MultiMek™ automated system
Unique diamond polished molding technology use no mold-releasing agents, minimizes wastage of precious samples and reagents while handling, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination