Axygen™ High Clarity Microplates†
Designed for use in sample collection, combinatorial chemistry, storage and HTS applications
$633.93 - $633.93
Specifications
Certifications/Compliance ANSI, SBS
For Use With (Application) Designed for use in sample collection, combinatorial chemistry, storage and HTS applications
Material Polypropylene
Type High-Clarity Microplate
Volume (Metric) Well 1.2 mL
View More Specs
Products 1
Catalog Number Mfr. No. Sterility Price Quantity  
Catalog Number Mfr. No. Sterility Price Quantity  
14-223-601 View Documents Corning Incorporated
PDW12HC
Non-sterile Case of 50 Each for $633.93
Description
Description
Made of virgin polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
  • 1.2mL volume per well
  • Ideal for sample collection, storage, and HTS
  • ANSI/SBS footprint for automation use
  • High clarity design allows visual confirmation of sample from bottom
  • 19 round well low-profile design for reduced space requirements
  • Ultra-flat surface ensures proper sealing of heat film
  • Centrifugation to 4000 RCF
  • Diamond polished wells allow complete sample recovery
Model P-DW-12-HC-S: Pre-sterilized
Specifications
Specifications
ANSI, SBS
Polypropylene
1.2 mL
Designed for use in sample collection, combinatorial chemistry, storage and HTS applications
High-Clarity Microplate