Axygen™ GentleFit™ Dual-Shot Filter Pipet Tips
Soft, leakproof seal provides a universal fit
$1,111.95 - $1,111.95
Specifications
Packaging Rack
Product Type Filter Tip
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
14-223-414 Corning Incorporated
TLF1000RS
1000μL Case of 960 Each for $1,111.95
Specifications
Description
These filter tips are manufactured from two different resins. The thermoplastic elastomer (TPE) top is easy to seal, provides a soft eject, and is colored-coded by volume. The rigid, polypropylene body ensures straightness, affords glass-like clarity and is non-reactive and graduated.
  • Versatile; dual-shot technology standardizes most research-grade pipetters
  • Available in three sizes: 10, 200 and 1000μL
  • Reduced sealing force
  • Minimal reciprocal ejection force
  • Economical; imposes less wear on pipetters
  • Ergonomic design; soft tip requires less pressure to create an optimum seal
  • Ideal for multichannel pipetters
  • Color-coded top identifies volume
  • Presterilized
  • Certified to be free from RNase, DNase, and human DNA
Description & Specifications
Specifications
Rack
Filter Tip