Axygen™ Gel Cutting Tips: 1.1 x 4mm
Fit most 1000uL pipettes. Available in bags and racks.
$117.19 - $416.68
Specifications
Certifications/Compliance RNase, DNase-free
Color Clear
Material Polypropylene
Sterility Nonsterile
Graduations 2, 5μL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Price Quantity  
Catalog Number Mfr. No. Packaging Price Quantity  
14-222-802 Corning Incorporated
TGL1140
250/bag; 10 bags/Cs.= 2500 Pack of 250 Each for $117.19
Case of 10 Pack for $1,019.00
14-222-803 Corning Incorporated
TGL1140R
48/rack; 10 racks/pk; 5 pks./Case = 2400 Pack of 480 Each for $416.68
Case of 5 Pack for $1,811.65
Specifications
Description
  • Universal fit for research grade pipetters
  • Assorted lengths to access bottom of narrow vessels and gel wells
  • 2 and 5μL graduations for quick volume check
Description & Specifications
Specifications
RNase, DNase-free
Polypropylene
2, 5μL
Clear
Nonsterile
Gel