missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Axygen™ FLIPR™ Liberty Robotic Tips
Accurate sample delivery
$2,186.40 - $2,186.40
Specifications
Clean Claims | DNase, RNase, and Pyrogen free |
---|---|
Autoclavable | Autoclavable |
For Use With (Equipment) | Molecular Devices™ FLIPR 384-Well Head |
Material | PP (Polypropylene) |
Tip Style | Automation |
Catalog Number | Mfr. No. | Color | Sterility | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Sterility | Price | Quantity | ||||
14-222-139
|
Axygen™
LT384BKR** |
Black | Non-sterile |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
14-222-140
|
Axygen™
LT384R |
Clear | Non-sterile |
Pack of 3840 Each for $2,186.40
Case of 5 Pack for $10,932.00 |
|
Description
- Available in clear and black
- Volume: 30μL
- Available as racked or racked and pre-sterilized
- For use with Molecular Devices Liberty head
- Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.
Certifications
Certified RNase-, DNase- and endotoxin-safe
Specifications
DNase, RNase, and Pyrogen free | |
Molecular Devices™ FLIPR 384-Well Head | |
Automation | |
50 Racks/Cs. |
Autoclavable | |
PP (Polypropylene) | |
30 μL | |
SBS Rack |