Axygen™ Caps for PCR Tubes
8-Strip domed or flat strip caps for Axygen PCR tubes
Supplier: Axygen™ PCR02CPY
Description
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
Specifications
RNase, DNase, pyrogen-free | |
Yellow | |
125/Pk. | |
8-strips PCR tubes and 96-well PCR microplates | |
Polypropylene | |
Strip |
Yellow | |
0.2 mL | |
Polypropylene | |
Non-sterile | |
Domed | |
8-Strip Domed PCR Tube Cap |