Axygen™ Biomek™ FX/NX Robotic Tips
Axygen robotic tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Supplier: Axygen™ FX384LR
Description
- Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
- Unsurpassed straightness to ensure tip enters the well smoothly, every time
- 384/1536-well compatible to meet your most demanding applications