missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ Beckman™ Biomek 1000 and 2000 Robotic Tips

Maxymum Recovery™ reduces sample loss and improves reproducibility and accuracy

$1,588.92 - $3,026.98

Specifications

Clean Claims DNase, Rnase Free
Color Clear
For Use With (Equipment) Beckman Coulter™ Biomek™ 1000/2000
Material PP (Polypropylene)
Shape Round
View More Specs

Products 2
Catalog Number Mfr. No. Filtered Sterility Volume (Metric) Tip Style Price Quantity  
Catalog Number Mfr. No. Filtered Sterility Volume (Metric) Tip Style Price Quantity  
50400339
View Documents
Peprotech
40023100UG
Non-filtered Non-sterile 50 μL Automation
Each for $1,588.92
 
50400340
View Documents
Peprotech
40023250UG
Non-filtered Sterile 50 μL Automation
Each for $3,026.98
 
Description

Description

Robotic tips uses unique diamond-polished molding technology use no mold-releasing agents. Tips minimize waste of precious samples and reagents. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.

  • Volume: 20 to 1000μL
  • Tip color: Clear
  • Rack color: Grey and Black
  • Available in Maxymum Recovery, Racked, Maxymum Recovery, Racked, Presterilized, Racked and Racked, Presterilized tip types

Order Info

Rack: 96 tips/rack, 10 racks/pack, 5 packs/case
Bulk Packed: 1000 tips/unit, 10 Units/case

BT-1000 series:
Rack: 72 tips/rack, 5 racks/case

Specifications

Specifications

DNase, Rnase Free
Beckman Coulter™ Biomek™ 1000/2000
Round
Clear
PP (Polypropylene)
SDS