Axygen™ Beckman™ Biomek 1000 and 2000 Robotic Tips
Maxymum Recovery™ reduces sample loss and improves reproducibility and accuracy
Manufacturer:  Corning Incorporated BT250R
 View more versions of this product
Catalog No. 14-222-053
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Order Info
Rack: 96 tips/rack, 10 racks/pack, 5 packs/case
Bulk Packed: 1000 tips/unit, 10 Units/case
BT-1000 series:
Rack: 72 tips/rack, 5 racks/case
Specifications
Specifications
Beckman Coulter™ Biomek™ 1000/2000
Automation
Non-Filtered
Clear
Round
Non-ANSI
Non-pyrogenic, uses unique diamond polished molding technology, use no mold-releasing agents, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination, minimize waste of precious samples and reagents
Rack
250μL
Non-sterile
Gray
PP (Polypropylene)
DNase, Rnase Free
Beckman Coulter™ Biomek™ 1000/2000
Documents