Axygen™ Axyprep™ Mini Spin Basket
Designed to be used with the AxyPrep 1.5 or 2mL capped microfuge tubes when capping during centrifugation is required
$2,099.13 - $2,099.13
Catalog Number | Mfr. No. | Capacity (Metric) | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Packaging | Price | Quantity | ||||
14-223-063
|
Corning Incorporated
APMCT15MLC |
1.5mL | 4000/Cs. |
Case of 4000 Each for $2,099.13
|
|
Description
An exceptionally useful device for many centrifuge-mediated sample prep applications. The internal volume is approximately 850mL and is excellent for applications such as gel filtration chromatography and nucleic acid purification. This spin basket can be modified by adding porous frits or retainer rings and membranes.
- Versatile design for many applications
- Mechanical filtration
- Gel filtration
- Silica membrane
- Affinity membrane
- Can be capped during centrifugation†
- Molded-in support grid
- Molded in medical grade polypropylene
- Polyethylene frits™ and retainer rings are sold separately
- Fits all common microfuge tubes