Please login to your online account to display your discounted pricing
Axygen™ Axyprep™ MAG Viral NA Kit
Designed for rapid and reliable isolation of DNA from whole blood, serum, plasma, saliva and other bodily fluids
$1,765.88 - $1,765.88
Specifications
For Use With (Application) For rapid and reliable isolation of DNA from whole blood, serum, plasma, saliva and other bodily fluids
Product Name MAG Viral NA Kit
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
14-223-425
corning incorporated
MAGVNAM
Medium; 384 Preps Case for $1,765.88
Description & Specifications
Specifications
For Use With (Application) For rapid and reliable isolation of DNA from whole blood, serum, plasma, saliva and other bodily fluids
Product Name MAG Viral NA Kit
High quality DNA is suitable for direct use in most downstream applications such as amplification and enzymatic reactions. The kit can be adapted to most major liquid handling workstations in the market.
Simplified extraction process
High quality and yield of nucleic acid purification
Manual and automation friendly
Compatible with genomic downstream applications: PCR, Sanger and next generation sequencing, genotyping and SNP detection, and gene expression