Please login to your online account to display your discounted pricing
Axygen™ AxyPrep™ MAG Soil, Stool and Water DNA Kit
One kit for several sample types - Ideal for environmental sample testing
$748.47 - $1,405.82
Specifications
For Use With (Application) DNA extraction and purification
Product Name AxyPrep Mag Soil, Stool and Water DNA Kit
View More Specs
Products
Catalog Number Mfr. No. Includes Sample Size Price Quantity    
14-223-416
corning incorporated
MAGSTLS
MAG-A1 Particles: 1.1; PTL Buffer: 60mL; Glass Beads: 15g; IR Reagent: 12mL; Binding Enhancer: 550μL; RNase A: 220μL; Pro K Solution: 1.1mL; Protocol Manual:1 Case for $748.47
14-223-417
corning incorporated
MAGSTLM
MAG-A1 Particles: 1.1; PTL Buffer: 2x300mL; Glass Beads: 50g; IR Reagent: 50mL; Binding Enhancer: 3mL; RNase A: 500μL; Pro K Solution: 5mL; Protocol Manual:1 Case for $1,405.82
Description & Specifications
Specifications
For Use With (Application) DNA extraction and purification
Product Name AxyPrep Mag Soil, Stool and Water DNA Kit
Allows rapid and reliable isolation of high-quality total DNA from fresh and frozen stool samples
Up to 200 mg stool samples can be processed in less than 60 minutes
Combines the reversible nucleic acid binding properties of MAG-A1 particles with the efficiency of IR Reagent to eliminate humic acid, polysaccharides, phenolic compounds, and enzyme inhibitors
Purified DNA is suitable for PCR, restriction digestion, and hybridization techniques
No organic extractions, which reduces plastic waste and hands-on time allowing multiple samples to be processed in parallel
  • Simplified DNA concentration process: 96 well microplate samples in less than an hour
  • Inhibitor-free DNA purification process
  • Compatible wtih genomics downstream applications
  • Manual and automation friendly
  • PCR and qPCR
  • Sequencing: Sanger and Next Generation
  • Genotyping and SNP detection
  • Restriction digestion