missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ AxyPrep Mag™ FragmentSelect Kits

Next generation sequencing sample preparation simplified

$316.59 - $4,792.90

Specifications

Format 96- and 384-well plate
For Use With (Application) Genomic DNA purification
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Quantity Price Quantity  
Catalog Number Mfr. No. Includes Quantity Price Quantity  
14-223-160 Corning Incorporated
MAGFRAGI5
AxyPrep™ Mag™ FragmentSelect-I paramagnetic bead solution 5mL Each for $316.59
14-223-161 Corning Incorporated
MAGFRAGI50
AxyPrep™ Mag™ FragmentSelect-I paramagnetic bead solution 50mL Each for $1,456.37
14-223-162 Corning Incorporated
MAGFRAGI250
AxyPrep™ Mag™ FragmentSelect-I paramagnetic bead solution 250mL Each for $4,792.90
Specifications

Description

Post DNA shearing, the library construction process for next generation sequencing mostly requires fragment selection regardless of the platform. Obtaining high recovery post fragment selection is becoming an important contributor for the reduction of sequencing bias. The AxyPrep FragmentSelect-I kit is optimized for Illumina next generation sequencing platforms and Life Technologies (SOLiD) fragment size selection needs, simplifying the library construction workflows. Where, the AxyPrep FragmentSelect-R is designed for fragment size selection needs in the Roche/454s DNA Genome sequencer library construction workflow. The AxyPrep Mag FragmentSelect kits utilize a unique paramagnetic bead technology for quick high-throughput optimized DNA size selection suitable for various next generation sequencing platforms.

  • Optimized fragment selection chemistries for Illumina, SOLiD and Roche/454
  • Versatile and flexible sequential fragment size selection: no gels needed
  • Efficient removal of primer-dimers; no need for further clean-up steps
  • Manual and automation friendly
  • Scalable: Tube, 96- and 384-well microplate formats
  • No centrifugation or filtration required
AxyPrep FragmentSelect kits consists of two major stages:
  • 1. Optimization of AxyPrep FragmentSelect beads ratio to input DNA for desired target DNA fragment size selection
  • 2. The optimal AxyPrep FragmentSelect beads ratio is utilized for the isolation of the desired target DNA fragment size
Note: AxyPrep FragmentSelect kits can be a one-stage process when large size DNA fragments are not a concern.

Description & Specifications

Specifications

96- and 384-well plate
Genomic DNA purification