Please login to your online account to display your discounted pricing

Axygen™ Axypet-16 Pipette Tips

For 1536-well pipetting

$1,640.05 - $6,501.46

Specifications

Certifications/Compliance RNase, DNase-free
Color Clear
Material Polypropylene
Product Type Tip
Volume (Metric) 0.5 to 30μL
View More Specs
Products
Catalog Number Mfr. No. Quantity Sterility Price Quantity    

14-223-373

 
corning incorporated
T1536LRS
19200 Case Sterile Pack of 3840 Each for $1,640.05
Case of 5 Pack for $7,130.65

50-400-903

 
corning incorporated
T1536LR
50 Case Nonsterile Case of 19200 Each for $6,501.46
Description & Specifications

Specifications

Certifications/Compliance RNase, DNase-free
Color Clear
Material Polypropylene
Product Type Tip
Volume (Metric) 0.5 to 30μL
For Use With (Application) For 1536-well pipetting
Packaging Rack
Product Type Tip

  • Tip Type: Maxymum Recovery
  • Tip Color: Clear
  • 0.5-30μL