Please login to your online account to display your discounted pricing

Axygen™ AxyMats™ Sealing Mats for 2mL 96 Well Plates with Square Wells

Minimize evaporation in wide range of temperatures in PCR and storage applications

$123.76 - $123.76

Specifications

Autoclavable Yes
Color Natural
Material Silicone
Well Shape Square
No. of Wells 96
View More Specs
Products
Catalog Number Mfr. No. Sterile Price Quantity    

14-222-012

 
corning incorporated
AM2MLSQ
No Pack of 10 Each for $123.76
Case of 5 Pack for $538.10
Description & Specifications

Specifications

Autoclavable Yes
Color Natural
Material Silicone
Well Shape Square
No. of Wells 96

For higher-throughput laboratories, Axygen offers sealing options to accommodate the need for greater automation with excellent cost-effectiveness. Each “well-sealer,” once punctured, will reseal itself, ensuring an airtight seal to the well. Used for PCR, compound storage, ELISA, tissue culture applications.

  • Made of research-grade silicone
  • Autoclavable mats can be re-used after bleach wash ethanol rinse
  • For use with 2mL 96 square well deep well plates