Axygen™ Aerosol Filter Tips, Sterile
Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation, comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Available in a wide variety of tip styles.
Manufacturer:  Axygen™ TF20LRS
 View more versions of this product
Catalog No. 14-222-781
Description
Description
Technologically advanced filter barrier pipette tips
  • Ideal for DNA amplification and microbiological applications where trace contamination can invalidate results
  • Filter allows free passage of air while preventing cross-contamination by blocking the passage of aerosols or liquids
  • Nonpyrogenic, clear polyethylene filters are free from DNA inhibitors and leachable additives
  • Every filter is inserted into the tip robotically to ensure freedom from nuclease contamination
  • Packaged in sealed-bottom racks with lids to prevent contamination
  • Wide variety of tip styles available in a range of packaging quantities:
    • Maxymum Recovery™ technology provides excellent accuracy and prevents wastage of precious samples
    • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
    • Other options include: universal fit, matrix and Eppendorf style
Specifications
Specifications
Gilson™ Pipetman™ P20
Maxymum Recovery
Filtered
Clear
USP 3127
Gilson™ Pipetman™ P20
Rack
0.5 to 20μL
Sterile
PP (Polypropylene)
DNase, RNase, and Pyrogen Free
Documents