Please login to your online account to display your discounted pricing
Axygen™ 96-Well PCR Microplates
Available in a variety of colors
$104.09 - $416.41
Specifications
Certifications/Compliance RNase, DNase-free
For Use With (Application) PCR thermal transfer, RT-PCR applications
Volume (Metric) Well 200μL
Well Color Clear
Material Polypropylene
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
14-221-902
corning incorporated
PCR96CS
Clear Pack of 10 Each for $104.09
Case of 5 Pack for $452.55
14-223-330
corning incorporated
PCR96A
Assorted Case of 50 Each for $315.97
50-400-620
corning incorporated
PCR96G
Green Case of 50 Each for $416.41
Description & Specifications
Specifications
Certifications/Compliance RNase, DNase-free
For Use With (Application) PCR thermal transfer, RT-PCR applications
Volume (Metric) Well 200μL
Well Color Clear
Material Polypropylene
Sterile No
PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications. A variety of colors are offered to help organize multi-step PCR processes.
  • 96-well PCR microplates
  • Working volume is 300μL
  • Available in a variety of colors
  • Sold 10 plates/unit; 5 units/Cs.