Please login to your online account to display your discounted pricing

Axygen™ 5μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 384 Tip Format

For use with Agilent/Velocity11 automation systems

$1,598.71 - $1,598.71

Specifications

Color Clear
For Use With (Equipment) Agilent™/Velocity11™ automated liquid handling system and VPrep™ and Bravo™ 384ST, 96ST and 16ST head
Material Polypropylene
Volume (Metric) 5μL
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
View More Specs
Products
Catalog Number Mfr. No. Certifications/Compliance Sterility Price Quantity    

14-222-892

 
corning incorporated
VT3845ULR
RNase, DNase-free, ANSI Nonsterile Pack of 3840 Each for $1,598.71
Case of 5 Pack for $6,950.90
Description & Specifications

Specifications

Color Clear
For Use With (Equipment) Agilent™/Velocity11™ automated liquid handling system and VPrep™ and Bravo™ 384ST, 96ST and 16ST head
Material Polypropylene
Volume (Metric) 5μL
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Quantity 384 Tips/Rack
Packaging Rack
Shape Round

Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

  • Clear polypropylene
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Available in nonsterile or sterile

Packaging:10 Racks/Pack, 5 Packs/Case

†With double label.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.