Axygen™ 384-Well PCR Half-Skirt Microplates
Optimized for MJ Research Cyclers
$698.23 - $698.23
Specifications
For Use With (Equipment) For ABI 3730, 3100 and MJ thermocyclers
Volume (Metric) Working 50μL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Color Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Color Price Quantity  
14-223-072 Corning Incorporated
PCR384M2CBC
For use within PCR thermal transfer and RT-PCR applications Clear Case of 50 Each for $698.23
Specifications
Description
PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications. A variety of colors are offered to help organize multi-step PCR processes.
  • 384-well PCR microplates
  • Working volume is 50μL
  • Optimized for MJ Research Cyclers
  • Available in a variety of colors
  • Compatible with ABI 3730/3100 and MJ Thermocyclers
  • Sold 10 plates/unit; 5 units/Cs.
Description & Specifications
Specifications
For ABI 3730, 3100 and MJ thermocyclers
50μL