missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ 300 μL Universal Pipetter Tips

Fine-point tips fits popular brands of pipettes

$88.48 - $985.17

Specifications

Volume (Metric) 1 to 300 μL
Color Clear
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Tip Style MicroPoint, Universal
For Use With (Equipment) Universal Fit
View More Specs

Products 3
Catalog Number Mfr. No. Sterility Format Price Quantity  
Catalog Number Mfr. No. Sterility Format Price Quantity  
14-222-746
View Documents Promotion Details
Axygen™
T350C
Non-sterile Bulk
Each for $88.48
Case of 10 Each for $884.80
 
14-222-751
View Documents Promotion Details
Axygen™
T350CRS
Sterile Rack
Case of 4800 Each for $985.17
 
14-222-750
View Documents Promotion Details
Axygen™
T350CR
Non-sterile Rack
Pack of 960 Each for $181.43
Case of 5 Pack for $907.15
 
Description

Description

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Fine-Point

Order Info

Bulk: 1000 tips/pack, 240 packs/case
Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

Specifications

Specifications

1 to 300 μL
DNase, RNase, ATP, and Endotoxin Free
Universal Fit
Clear
MicroPoint, Universal
Polypropylene
SDS
missing translation for 'documents'

missing translation for 'documents'