Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Beveled, Reference
Designed to fit a wide variety of single and multi-channel pipettes
Supplier:  Axygen™ TR222YR
 View more versions of this product
Catalog No. 50-400-997
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
Specifications
Specifications
1 to 200 μL
Non-sterile
Autoclavable
Rack
Hinged rack
Yes
10, 50, 100μL
Non-Filtered
Yellow
DNase, RNase, and Pyrogen free
Beveled, Universal
Universal Fit
Polypropylene