Please login to your online account to display your discounted pricing
Axygen™ 1200μL Universal Pipette Tips
Fits most popular brands of pipettes
$48.81 - $48.81
Products
Catalog Number Mfr. No. Price Quantity    
14-222-705
corning incorporated
T1200C
Pack of 1000 Each for $48.81
Case of 5 Pack for $212.20
Description & Specifications
  • Optimized tip bevel reduces sample hang-up and improves pipetting accuracy
  • Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
  • Volume range: 100 to 1200μL
  • Available in bulk or racked formats