Axygen™ 1000μL Universal Pipette Tips: Wide Bore, Maxymum Recovery
Compatible with most popular brands of pipettes
$559.51 - $559.51
Specifications
Autoclavable Yes
For Use With (Equipment) Single and multichannel pipettors
Material Polypropylene
Volume (Metric) 100 to 1000μL
Tip Style Universal Fit
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Certifications/Compliance Color Packaging Sterility Price Quantity  
Catalog Number Mfr. No. Certifications/Compliance Color Packaging Sterility Price Quantity  
14-222-700 Corning Incorporated
T1005WBCLR
DNA, DNase, RNase and PCR inhibitor-free Clear Rack Nonsterile Case of 1000 Each for $559.51
Specifications
Description
  • Maxymum Recovery™ tips allows complete retrieval of precious samples
  • Orifice size of 0.065 in. (1.6mm) makes wide bore tips suitable for pipetting viscous materials, genomic DNA or delicate cell culturese
  • Volume range: 100 to 1000μL
  • Available in racked format
Description & Specifications
Specifications
Yes
Polypropylene
Universal Fit
Single and multichannel pipettors
100 to 1000μL