Axygen™ 10µL Microvolume Filter Tips
Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, bevelled, Eppendorf-style, nonsterile, sterile, non-filtered, and other features for specialized applications.
Manufacturer:  Axygen™ TF300RS
 View more versions of this product
Catalog No. 14-222-785
Description
Description
  • Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Microvolume
  • Maxymum Recovery technology for ultra-low retention and high yield
Specifications
Specifications
Gilson™ Pipetman™ P10,Gilson™ Pipetman™ P2
Microvolume,Universal Fit
Sterile
PP (Polypropylene)
DNase, Rnase Free
Gilson™ Pipetman™ P10
Rack
0.5 to 10μL
Clear
USP 3127
Allows recovery of costly samples with improved precision and accuracy
Documents